missing translation for 'onlineSavingsMsg'
Learn More

Triethoxyphenylsilane 99.0+%, TCI America™

Catalog No. P073525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P073525ML 25 mL
P0735100ML 100 mL
P0735500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P073525ML Supplier TCI America Supplier No. P073525ML

Chemical Identifiers

CAS 780-69-8
Molecular Formula C12H20O3Si
Molecular Weight (g/mol) 240.37
MDL Number MFCD00009065
InChI Key JCVQKRGIASEUKR-UHFFFAOYSA-N
Synonym phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15
PubChem CID 13075
IUPAC Name triethoxy(phenyl)silane
SMILES CCO[Si](OCC)(OCC)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 235°C
Quantity 25 mL
Formula Weight 240.37
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxyphenylsilane
EINECSNumber (3)-2635
RTECSNumber VV4900000
TSCA Yes