missing translation for 'onlineSavingsMsg'
Learn More

Triethoxymethylsilane 98.0+%, TCI America™

Catalog No. p-7132179
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045125ML 25 mL
M0451500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M045125ML Supplier TCI America Supplier No. M045125ML
Only null left

Chemical Identifiers

CAS 2031-67-6
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.30
MDL Number MFCD00009066
InChI Key CPUDPFPXCZDNGI-UHFFFAOYSA-N
Synonym methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes
PubChem CID 16241
IUPAC Name triethoxy(methyl)silane
SMILES CCO[Si](C)(OCC)OCC

Specifications

Color Colorless
Boiling Point 143°C
Quantity 25 mL
UN Number 1993
Formula Weight 178.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxymethylsilane
EINECSNumber (2)-2052&(2)-2053
RTECSNumber VV4640000
TSCA Yes