missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethoxy(propyl)silane 98.0+%, TCI America™
$41.02 - $102.52
Chemical Identifiers
| CAS | 2550-02-9 |
|---|---|
| Molecular Formula | C9H22O3Si |
| Molecular Weight (g/mol) | 206.36 |
| MDL Number | MFCD00026756 |
| InChI Key | NBXZNTLFQLUFES-UHFFFAOYSA-N |
| Synonym | propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane |
| PubChem CID | 75691 |
| IUPAC Name | triethoxy(propyl)silane |
| SMILES | CCC[Si](OCC)(OCC)OCC |
Chemical Identifiers
| 2550-02-9 | |
| 206.36 | |
| NBXZNTLFQLUFES-UHFFFAOYSA-N | |
| 75691 | |
| CCC[Si](OCC)(OCC)OCC |
| C9H22O3Si | |
| MFCD00026756 | |
| propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane | |
| triethoxy(propyl)silane |
Specifications
| 2550-02-9 | |
| 180°C | |
| MFCD00026756 | |
| 1993 | |
| NBXZNTLFQLUFES-UHFFFAOYSA-N | |
| triethoxy(propyl)silane | |
| 75691 | |
| ≥98.0% (GC) | |
| Triethoxy(propyl)silane |
| Colorless | |
| C9H22O3Si | |
| 25 g | |
| propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane | |
| CCC[Si](OCC)(OCC)OCC | |
| 206.36 | |
| 206.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2052&(2)-2053
TSCA : Yes