missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy(propyl)silane 98.0+%, TCI America™

Catalog No. T286725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T286725G 25 g
T2867100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T286725G Supplier TCI America Supplier No. T286725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2550-02-9
Molecular Formula C9H22O3Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00026756
InChI Key NBXZNTLFQLUFES-UHFFFAOYSA-N
Synonym propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane
PubChem CID 75691
IUPAC Name triethoxy(propyl)silane
SMILES CCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 180°C
Quantity 25 g
UN Number 1993
Formula Weight 206.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy(propyl)silane
EINECSNumber (2)-2052&(2)-2053
TSCA Yes