missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethoxy(pentafluorophenyl)silane 95.0+%, TCI America™
$109.98 - $329.94
Chemical Identifiers
| CAS | 20083-34-5 |
|---|---|
| Molecular Formula | C12H15F5O3Si |
| Molecular Weight (g/mol) | 330.326 |
| MDL Number | MFCD03411257 |
| InChI Key | QALDFNLNVLQDSP-UHFFFAOYSA-N |
| PubChem CID | 2782625 |
| IUPAC Name | triethoxy-(2,3,4,5,6-pentafluorophenyl)silane |
| SMILES | CCO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC |
Chemical Identifiers
| 20083-34-5 | |
| 330.326 | |
| QALDFNLNVLQDSP-UHFFFAOYSA-N | |
| triethoxy-(2,3,4,5,6-pentafluorophenyl)silane |
| C12H15F5O3Si | |
| MFCD03411257 | |
| 2782625 | |
| CCO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC |
Specifications
| 20083-34-5 | |
| 238°C | |
| MFCD03411257 | |
| QALDFNLNVLQDSP-UHFFFAOYSA-N | |
| triethoxy-(2,3,4,5,6-pentafluorophenyl)silane | |
| 2782625 | |
| ≥95.0% (GC) | |
| Triethoxy(pentafluorophenyl)silane |
| Yellow | |
| C12H15F5O3Si | |
| 1 g | |
| CCO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC | |
| 330.326 | |
| 330.33 | |
| Liquid |
Safety and Handling
TSCA : No