missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy-n-octylsilane 97.0+%, TCI America™

Catalog No. p-7134221
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
O017125ML 25 mL
O0171100ML 100 mL
O0171500ML 500 mL
3 options

Catalog No. O017125ML

Supplier: TCI America O017125ML

Only null left

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 106°C
Quantity 25 mL
Formula Weight 276.49
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy-n-octylsilane
EINECSNumber (2)-3784
TSCA Yes