missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane 90.0+%, TCI America™

Catalog No. T32461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32461G 1 g
T32465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32461G Supplier TCI America Supplier No. T32461G

Chemical Identifiers

CAS 130676-81-2
Molecular Formula C15H21F13O3Si
Molecular Weight (g/mol) 524.39
MDL Number MFCD27918526
InChI Key JVAFDQZZUMUFSM-UHFFFAOYSA-N
PubChem CID 14794618
IUPAC Name triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane
SMILES CCO[Si](CCCC(C(F)(F)F)(C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC

Specifications

Color Colorless
Boiling Point 81°C
Quantity 1 g
Formula Weight 524.39
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane