missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Technical, 85%, Spectrum™ Chemical

Catalog No. p-7166686
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
20 L
200 L
4 L
500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Catalog No. Quantity Packaging
18614543 500 mL Amber Glass Bottle
18614545 4 L Amber Glass Bottle
18614544 20 L Poly Drum
18614542 200 L Drum
4 options

Catalog No. 18614543

Supplier: Spectrum Chemical Mfg Cor T2561500MLGL

Only null left

Chemical Identifiers

CAS 102-71-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 0.85
Grade Technical

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.