missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine, Technical, 85%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 335°C |
| Packaging | Drum |
| Quantity | 4 L |
| Molecular Weight (g/mol) | 149.19 |
| Percent Purity | 0.85 |
| Grade | Technical |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Triethanolamine |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.