missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Reagent, 99-107.4%, Spectrum™ Chemical

Catalog No. 18607564
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
20 L
4 x 4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607564 20 L Amber Glass Bottle
18607561 6 x 500 mL Amber Glass Bottle
18607562 4 L Amber Glass Bottle
18607563 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18607564 Supplier Spectrum Chemical Mfg Cor Supplier No. T256420LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T2564, 102-71-6, C6H15NO3

Spectrum™ Chemical Triethanolamine, Reagent is a is colorless organic compound that has a viscous texture and is both a tertiary amine and a triol. It is mainly used as an emulsifier and a surfactant. It neutralizes fatty acids while adjusting and buffering the pH. It is used in the fabrication of cosmetics and medicine such as ear drops, skin lotions, eye gels, shampoos and others. It also has a use in the laboratory as a complexing agent for aluminum ions in aqueous solutions. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

CAS Max % 1
Boiling Point 335°C
Packaging Amber Glass Bottle
Quantity 20 L
Percent Purity 99.0 to 107.4%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Triethanolamine