missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine Phosphate 98.0+%, TCI America™
Supplier: TCI America N0103500G
Specifications
| Triethanolamine Phosphate | |
| 106°C | |
| C6H18NO7P | |
| 500 g | |
| NHFDKKSSQWCEES-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethanol;phosphoric acid | |
| 61430 | |
| ≥98.0% (T) |
| 10017-56-8 | |
| White | |
| MFCD00054303 | |
| 2,2′,2′′-Nitrilotriethanol Phosphate | |
| C(CO)N(CCO)CCO.OP(=O)(O)O | |
| 247.184 | |
| 247.18 | |
| Crystalline Powder |
Chemical Identifiers
| 10017-56-8 | |
| 247.184 | |
| NHFDKKSSQWCEES-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethanol;phosphoric acid |
| C6H18NO7P | |
| MFCD00054303 | |
| 61430 | |
| C(CO)N(CCO)CCO.OP(=O)(O)O |
Safety and Handling
TSCA : Yes