missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine Phosphate 98.0+%, TCI America™

Catalog No. N010325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010325G 25 g
N0103500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N010325G Supplier TCI America Supplier No. N010325G

Chemical Identifiers

CAS 10017-56-8
Molecular Formula C6H18NO7P
Molecular Weight (g/mol) 247.184
MDL Number MFCD00054303
InChI Key NHFDKKSSQWCEES-UHFFFAOYSA-N
PubChem CID 61430
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol;phosphoric acid
SMILES C(CO)N(CCO)CCO.OP(=O)(O)O

Specifications

Melting Point 106°C
Color White
Quantity 25 g
Synonym 2,2′C,2′C′C-Nitrilotriethanol Phosphate
Formula Weight 247.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethanolamine Phosphate