Learn More
Triethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals
$106.97 - $1496.97
Chemical Identifiers
| CAS | 637-39-8 |
|---|---|
| Molecular Formula | C6H16ClNO3 |
| Molecular Weight (g/mol) | 185.648 |
| MDL Number | MFCD00012596 |
| InChI Key | HHLJUSLZGFYWKW-UHFFFAOYSA-N |
| Synonym | triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy |
| PubChem CID | 101814 |
| IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride |
| SMILES | C(CO)N(CCO)CCO.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1567830
|
Thermo Scientific Chemicals
A1567830 |
250 g |
Each for $106.97
|
|
|||||
|
AAA156780B
|
Thermo Scientific Chemicals
A156780B |
1000 g |
Each for $298.34
|
|
|||||
|
AAA156780I
|
Thermo Scientific Chemicals
A156780I |
5000 g |
Each for $1,496.97
|
|
|||||
Description
Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsProtein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
| 637-39-8 | |
| 185.648 | |
| HHLJUSLZGFYWKW-UHFFFAOYSA-N | |
| 101814 | |
| C(CO)N(CCO)CCO.Cl |
| C6H16ClNO3 | |
| MFCD00012596 | |
| triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy | |
| 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride |
Specifications
| 637-39-8 | |
| C6H16ClNO3 | |
| MFCD00012596 | |
| 3909940 | |
| 14,9665 | |
| Soluble in water. | |
| C(CO)N(CCO)CCO.Cl | |
| 185.648 | |
| 185.65 | |
| Triethanolamine hydrochloride |
| 177°C to 179°C | |
| (HOCH2CH2)3N·HCl | |
| 250 g | |
| Hygroscopic | |
| triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy | |
| HHLJUSLZGFYWKW-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride | |
| 101814 | |
| ≥99% |
Safety and Handling
EINECSNumber : 211-284-2
RTECSNumber : KL9346500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only