missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine (Certified), Fisher Chemical™

Catalog No. p-203471
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T4071 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T4071 Supplier Thermo Fisher Scientific Supplier No. T407 1
Only null left

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO

Specifications

Decomposition Information >325°C
Melting Point 21°C
Color Yellow
Density 1.125g/cm³
pH 10.5
Boiling Point 360°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Linear Formula N(CH2CH2OH)3
Quantity 1 L
Vapor Pressure <0.01mmHg at 20°C
Formula Weight 149.14
Percent Purity ≥99%
Grade Certified
Physical Form Viscous Liquid
Chemical Name or Material Triethanolamine
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0