missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine Borate 95.0+%, TCI America™
Supplier: TCI America T1291500G
Specifications
| Triethanolamine Borate | |
| 237°C | |
| C6H12BNO3 | |
| 500 g | |
| NKPKVNRBHXOADG-UHFFFAOYSA-N | |
| 4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane | |
| 84862 | |
| ≥95.0% (T) |
| 283-56-7 | |
| White | |
| MFCD00003272 | |
| 2,2′,2′′-Nitrilotriethyl Borate | |
| B12OCCN(CCO1)CCO2 | |
| 156.976 | |
| 156.98 | |
| Crystalline Powder |
Chemical Identifiers
| 283-56-7 | |
| 156.976 | |
| NKPKVNRBHXOADG-UHFFFAOYSA-N | |
| 4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane |
| C6H12BNO3 | |
| MFCD00003272 | |
| 84862 | |
| B12OCCN(CCO1)CCO2 |
Safety and Handling
EINECSNumber : (2)-2100
RTECSNumber : YJ8968050
TSCA : No