missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine, BAKER ANALYZED™ Reagent, J.T. Baker™
Specifications
Specifications
| Flash Point | 179.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Quantity | 500 mL |
| Specific Gravity | 1L = 1.13kg |
| Formula Weight | 149.19 |
| Percent Purity | ≥99.0% |
| Grade | Not Available |
| Chemical Name or Material | Triethanolamine |