missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 97%, Thermo Scientific Chemicals

Catalog No. p-203763
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC139560010 1 L
AC139560025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139560010 Supplier Thermo Scientific Chemicals Supplier No. 139560010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-71-6
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621

Specifications

Decomposition Information >325°C
Melting Point 21.0°C
Density 1.1245g/mL
Boiling Point 360.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H15NO3
Refractive Index 1.4840 to 1.4860
Linear Formula (HOCH2CH2)3N
Quantity 1 L
Fieser 01,1196
Specific Gravity 1.1245
Merck Index 15, 9830
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohol, methanol and acetone, soluble in chloroform, slichtly soluble in benzene and ether, soluble 4.2% in benzene; 1.6% in ether; 0.4% in, carbon tetrachloride and < 0.1% in n-hepta
SMILES C(CO)N(CCO)CCO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
Vapor Pressure <0.01mmHg at 20°C
Formula Weight 149.19
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material Triethanolamine, 97%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-049-8
RTECSNumber KL9275000
TSCA TSCA

RUO – Research Use Only