missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine (2,2,2-Nitrilotriethanol), Reagents

Catalog No. p-200020960
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336911 4 L Poly Bottle
01336910 4 L Glass Bottle
01336909 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336911 Supplier Labchem Inc Supplier No. C23425004A
Only null left

Triethanolamine (2,2,2-Nitrilotriethanol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym 2,2,2-Nitrilotriethanol, Trolamine
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Boiling Range 335°C
Melting Point 21°C
Color Colorless to Slight Yellow
Boiling Point 335°C
Melting Range 21°C
Packaging Poly Bottle
Quantity 4 L
Specific Gravity 1.1242
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triethanolamine (2,2,2-Nitrilotriethanol)
Show More Show Less