Learn More
New England Biolab Ltd TriDye™ 1 kb Plus DNA Ladder 1.25 ml
Supplier:  New England Biolab Ltd N3270S
Encompass
TriDye 1 kb Plus DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference points.
Catalog No. 50811781
Only null left