missing translation for 'onlineSavingsMsg'
Learn More

Tridodecylamine, 90%

Catalog No. p-203422
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421620250 25 g Glass bottle
AC421621000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421620250 Supplier Thermo Scientific Chemicals Supplier No. 421620250

CAS: 102-87-4 | C36H75N | 522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-87-4
Molecular Formula C36H75N
Molecular Weight (g/mol) 522
MDL Number MFCD00008971
InChI Key SWZDQOUHBYYPJD-UHFFFAOYSA-N
Synonym tridodecylamine, trilaurylamine, tri-n-dodecylamine, alamine 304, tridodecyl amine, adogen 360, 1-dodecanamine, n,n-didodecyl, n,n-didodceyl-1-dodecanamine, n,n-didodecyl-1-dodecanamine, unii-sf36t21u17
PubChem CID 7624
IUPAC Name N,N-didodecyldodecan-1-amine
SMILES CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

Specifications

Melting Point 16°C
Color Yellow
Density 0.8200g/mL
Boiling Point 450°C
Flash Point 216°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Refractive Index 1.4568 to 1.4588
Linear Formula (CH3(CH2)11)3N
Quantity 25 g
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble
Formula Weight 522
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Tridodecylamine, 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 203-063-4
TSCA TSCA

RUO – Research Use Only