missing translation for 'onlineSavingsMsg'
Learn More

Tridecanoic Acid 98.0+%, TCI America™

Catalog No. T041225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T041225G 25 g
T0412500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T041225G Supplier TCI America Supplier No. T041225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-53-9
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00002741
InChI Key SZHOJFHSIKHZHA-UHFFFAOYSA-N
Synonym n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16
PubChem CID 12530
ChEBI CHEBI:45919
IUPAC Name tridecanoic acid
SMILES CCCCCCCCCCCCC(=O)O

Specifications

Melting Point 41°C
Color White
Boiling Point 236°C
Quantity 25 g
Formula Weight 214.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tridecanoic Acid
EINECSNumber (2)-0608
RTECSNumber YD3850000
TSCA Yes