missing translation for 'onlineSavingsMsg'
Learn More

Tridecane 99.0+%, TCI America™

Catalog No. T041125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T041125ML 25 mL
T0411100ML 100 mL
T0411500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T041125ML Supplier TCI America Supplier No. T041125ML

Chemical Identifiers

CAS 629-50-5
Molecular Formula C13H28
Molecular Weight (g/mol) 184.37
MDL Number MFCD00008979
InChI Key IIYFAKIEWZDVMP-UHFFFAOYSA-N
Synonym n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
PubChem CID 12388
ChEBI CHEBI:35998
IUPAC Name tridecane
SMILES CCCCCCCCCCCCC

Specifications

Melting Point -5°C
Color Colorless
Boiling Point 235°C
Quantity 25 mL
Formula Weight 184.37
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecane
EINECSNumber (2)-0010
RTECSNumber YD3025000
TSCA Yes