missing translation for 'onlineSavingsMsg'
Learn More

Tridecanal, 96%, stab.

Catalog No. p-7024661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1972206 5 g
AAA1972214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1972206 Supplier Thermo Scientific Chemicals Supplier No. A1972206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10486-19-8 | C13H26O | 198.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10486-19-8
Molecular Formula C13H26O
Molecular Weight (g/mol) 198.35
MDL Number MFCD00007018
InChI Key BGEHHAVMRVXCGR-UHFFFAOYSA-N
Synonym tridecyl aldehyde, tridecanaldehyde, 1-tridecanal, n-tridecylaldehyde, n-tridecanal, tridecane aldehyde, tridecanealdehyde, aldehyde c-13, technical, tridecanal
PubChem CID 25311
IUPAC Name tridecanal
SMILES CCCCCCCCCCCCC=O

Specifications

Melting Point 14°C to 15°C
Density 0.83
Boiling Point 280°C
Flash Point 132°C (269°F)
Odor Characteristic
Assay Percent Range 0.94
Refractive Index 1.4384
Linear Formula CH3(CH2)11CHO
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 198.35
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tridecanal, stabilized
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 234-004-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only