missing translation for 'onlineSavingsMsg'
Learn More

Tridecafluorohexyl Bromide 97.0+%, TCI America™

Catalog No. T24795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24795G 5 g
T247925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24795G Supplier TCI America Supplier No. T24795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 335-56-8
Molecular Formula C6BrF13
Molecular Weight (g/mol) 398.949
MDL Number MFCD00042349
InChI Key JTYRBFORUCBNHJ-UHFFFAOYSA-N
Synonym 1-bromoperfluorohexane, perfluorohexyl bromide, perfluorohexylbromide, tridecafluorohexyl bromide, 1-bromo-tridecafluorohexane, bromoperfluorohexane, acmc-1cmhf, 1-bromotridecafluorohexane, tridecafluoro-1-bromohexane
PubChem CID 92755
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
SMILES C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 398.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecafluorohexyl Bromide