missing translation for 'onlineSavingsMsg'
Learn More

Tridecafluoroheptanoic Acid High Grade 98.0+%, TCI America™

Catalog No. A57215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57215G 5 g
A57211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A57215G Supplier TCI America Supplier No. A57215G

Chemical Identifiers

CAS 375-85-9
Molecular Formula C7HF13O2
Molecular Weight (g/mol) 364.062
MDL Number MFCD00039604
InChI Key ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
Synonym perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
PubChem CID 67818
ChEBI CHEBI:35547
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
SMILES C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Color White
Boiling Point 177°C
Quantity 5 g
UN Number 3261
Formula Weight 364.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tridecafluoroheptanoic Acid High Grade [Ion-Pair Reagent for LC-MS]
EINECSNumber (2)-1182&(2)-2659
TSCA Yes