missing translation for 'onlineSavingsMsg'
Learn More

Tridecafluoroheptanoic Acid (ca. 5mmol) 98.0+%, TCI America™

Catalog No. A5716SET
Change view
Click to view available options
Quantity:
Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5716SET Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5716SET Supplier TCI America Supplier No. A57161SAMPLE

Chemical Identifiers

CAS 375-85-9
Molecular Formula C7HF13O2
Molecular Weight (g/mol) 364.062
MDL Number MFCD00039604
InChI Key ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
Synonym perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
PubChem CID 67818
ChEBI CHEBI:35547
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
SMILES C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Color White-Yellow
Boiling Point 177°C
Quantity Set
UN Number 3261
Formula Weight 364.06
Percent Purity ≥98.0% (T)
Physical Form Crystals
Chemical Name or Material Tridecafluoroheptanoic Acid (ca. 5mmol) [Ion-Pair Reagent for LC-MS]
EINECSNumber (2)-1182&(2)-2659
TSCA Yes