missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexyltin chloride, 97%

Catalog No. p-7036983
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5549303 1 g
AAH5549314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5549303 Supplier Thermo Scientific Chemicals Supplier No. H5549303
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 3091-32-5 | C18H33ClSn | 403.622 g/mol

Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3091-32-5
Molecular Formula C18H33ClSn
Molecular Weight (g/mol) 403.622
MDL Number MFCD00003817
InChI Key OJVZYIKTLISAIH-UHFFFAOYSA-M
Synonym tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin
PubChem CID 76539
IUPAC Name chloro(tricyclohexyl)stannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl

Specifications

Melting Point 123°C to 125°C
Linear Formula (C6H11)3SnCl
Quantity 1 g
UN Number UN3146
Solubility Information Soluble in ethanol,ether,toluene and chloroform. Insoluble in water.
Formula Weight 403.6
Percent Purity 97%
Chemical Name or Material Tricyclohexyltin chloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 221-437-5
RTECSNumber WH6830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only