missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine, technical grade

Catalog No. p-7059558
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4485403 1 g
AA4485406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4485403 Supplier Thermo Scientific Chemicals Supplier No. 04485403
Only null left

CAS: 2622-14-2 | C18H33P | 280.436 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2622-14-2
Molecular Formula C18H33P
Molecular Weight (g/mol) 280.436
MDL Number MFCD00003853
InChI Key WLPUWLXVBWGYMZ-UHFFFAOYSA-N
Synonym tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
PubChem CID 75806
IUPAC Name tricyclohexylphosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

Specifications

Melting Point 76°C to 78°C
Color White
Odor Unpleasant
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 280.42
Grade Technical
Physical Form Solid
Chemical Name or Material Tricyclohexylphosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-069-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only