missing translation for 'onlineSavingsMsg'
Learn More

Tricresyl Phosphate (mixture of isomers) 99.0+%, TCI America™

Catalog No. p-7134528
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P027325G 25 g
P0273500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P027325G Supplier TCI America Supplier No. P027325G
Only null left

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
MDL Number MFCD02102116
Synonym Phosphoric Acid Tritolyl Ester, Phosphoric Acid Tricresyl Ester, Tritolyl Phosphate

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 255°C
Quantity 25 g
UN Number 3082
Formula Weight 368.37
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Tricresyl Phosphate (mixture of isomers)
EINECSNumber (3)-2522&(3)-2613&(3)-3363
RTECSNumber TD0175000
TSCA Yes