missing translation for 'onlineSavingsMsg'
Learn More

Tricosanol, 99+%

Catalog No. AC258161000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC258161000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC258161000 Supplier Thermo Scientific Chemicals Supplier No. 258161000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3133-01-5 | C23H48O | 340.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3133-01-5
Molecular Formula C23H48O
Molecular Weight (g/mol) 340.64
MDL Number MFCD00044484
InChI Key FPLNRAYTBIFSFW-UHFFFAOYSA-N
Synonym 1-tricosanol, tricosanol, tricosyl alcohol, n-tricosanol, unii-k22080opox, n-tricosyl alcohol, acmc-2097cw, 1-tricosanol 10 microg/ml in methyl-tert. butyl ether
PubChem CID 18431
ChEBI CHEBI:78411
IUPAC Name tricosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 74°C to 76°C
Color White
Boiling Point 185°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)22OH
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ethers,ketones,specially when hot
Formula Weight 340.63
Percent Purity 99+%
Physical Form Fine Crystalline Powder or Flakes
Chemical Name or Material Tricosanol, 99+%
Show More Show Less
EINECSNumber 221-527-4

RUO – Research Use Only