missing translation for 'onlineSavingsMsg'
Learn More

Tricosane 98.0+%, TCI America™

Supplier:  TCI America T056810G

Catalog No. T056810G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tricosane
47°C
234°C
MFCD00009350
n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
CCCCCCCCCCCCCCCCCCCCCCC
324.637
CHEBI:32934
≥98.0% (GC)
638-67-5
White
C23H48
10 g
FIGVVZUWCLSUEI-UHFFFAOYSA-N
tricosane
12534
324.64
Crystalline Powder

Chemical Identifiers

638-67-5
324.637
FIGVVZUWCLSUEI-UHFFFAOYSA-N
12534
tricosane
C23H48
MFCD00009350
n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
CHEBI:32934
CCCCCCCCCCCCCCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0010

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions