missing translation for 'onlineSavingsMsg'
Learn More

Tricosane, 98%

Catalog No. AC421600010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421600010 1 g Glass bottle
AC421600050 5 g Glass bottle
AC421600250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC421600010 Supplier Thermo Scientific Chemicals Supplier No. 421600010

CAS: 638-67-5 | C23H48 | 324.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-67-5
Molecular Formula C23H48
Molecular Weight (g/mol) 324.63
MDL Number MFCD00009350
InChI Key FIGVVZUWCLSUEI-UHFFFAOYSA-N
Synonym n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
PubChem CID 12534
ChEBI CHEBI:32934
IUPAC Name tricosane
SMILES CCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 47°C to 50°C
Color White
Boiling Point 380°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)21CH3
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 324.63
Percent Purity 98%
Physical Form Shiny Flakes
Chemical Name or Material Tricosane
Show More Show Less
EINECSNumber 211-347-4

RUO – Research Use Only