missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricosafluorododecanoic Acid 92.0+%, TCI America™
$65.46 - $229.10
Chemical Identifiers
| CAS | 307-55-1 |
|---|---|
| Molecular Formula | C12HF23O2 |
| Molecular Weight (g/mol) | 614.101 |
| MDL Number | MFCD00198081 |
| InChI Key | CXGONMQFMIYUJR-UHFFFAOYSA-N |
| Synonym | perfluorododecanoic acid, tricosafluorododecanoic acid, perfluorolauric acid, dodecanoic acid, tricosafluoro, unii-p6g1j7nm2d, p6g1j7nm2d, pfdoa, tricosafluorolauric acid, n-perfluorododecanoic acid, n-c11f23cooh |
| PubChem CID | 67545 |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid |
| SMILES | C(=O)(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
Chemical Identifiers
| 307-55-1 | |
| 614.101 | |
| CXGONMQFMIYUJR-UHFFFAOYSA-N | |
| 67545 | |
| C(=O)(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| C12HF23O2 | |
| MFCD00198081 | |
| perfluorododecanoic acid, tricosafluorododecanoic acid, perfluorolauric acid, dodecanoic acid, tricosafluoro, unii-p6g1j7nm2d, p6g1j7nm2d, pfdoa, tricosafluorolauric acid, n-perfluorododecanoic acid, n-c11f23cooh | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid |
Specifications
| 307-55-1 | |
| White | |
| C12HF23O2 | |
| 1 g | |
| perfluorododecanoic acid, tricosafluorododecanoic acid, perfluorolauric acid, dodecanoic acid, tricosafluoro, unii-p6g1j7nm2d, p6g1j7nm2d, pfdoa, tricosafluorolauric acid, n-perfluorododecanoic acid, n-c11f23cooh | |
| C(=O)(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
| 614.101 | |
| 614.10 | |
| Crystalline Powder |
| 110°C | |
| 245°C | |
| MFCD00198081 | |
| 3261 | |
| CXGONMQFMIYUJR-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid | |
| 67545 | |
| ≥92.0% (GC) | |
| Tricosafluorododecanoic Acid |
Safety and Handling
EINECSNumber : (2)-2658&(2)-2659
RTECSNumber : JR3740000
TSCA : Yes