missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricosafluorododecanoic Acid 92.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | White |
| Boiling Point | 245°C |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 614.10 |
| Percent Purity | ≥92.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tricosafluorododecanoic Acid |
Safety and Handling
| EINECSNumber | (2)-2658&(2)-2659 |
| RTECSNumber | JR3740000 |
| TSCA | Yes |