missing translation for 'onlineSavingsMsg'
Learn More

Tricosafluorododecanoic Acid 92.0+%, TCI America™

Catalog No. T24921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24921G 1 g
T24925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24921G Supplier TCI America Supplier No. T24921G
Only null left

Chemical Identifiers

CAS 307-55-1
Molecular Formula C12HF23O2
Molecular Weight (g/mol) 614.101
MDL Number MFCD00198081
InChI Key CXGONMQFMIYUJR-UHFFFAOYSA-N
Synonym perfluorododecanoic acid, tricosafluorododecanoic acid, perfluorolauric acid, dodecanoic acid, tricosafluoro, unii-p6g1j7nm2d, p6g1j7nm2d, pfdoa, tricosafluorolauric acid, n-perfluorododecanoic acid, n-c11f23cooh
PubChem CID 67545
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 110°C
Color White
Boiling Point 245°C
Quantity 1 g
UN Number 3261
Formula Weight 614.10
Percent Purity ≥92.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tricosafluorododecanoic Acid
EINECSNumber (2)-2658&(2)-2659
RTECSNumber JR3740000
TSCA Yes