missing translation for 'onlineSavingsMsg'
Learn More

Tricine 99.0+%, TCI America™

Catalog No. p-7136394
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T068225G 25 g
T0682250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T068225G Supplier TCI America Supplier No. T068225G
Only null left

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym tricine, n-tris hydroxymethyl methylglycine, n-tri hydroxymethyl methyl glycine, n-tris hydroxymethyl methyl glycine, glycine, n-2-hydroxy-1,1-bis hydroxymethyl ethyl, n-2-hydroxy-1,1-bis hydroxymethyl ethyl glycine, unii-w12lh4v8v3, n-tris-hydroxymethyl methylglycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Color White
Quantity 25 g
Formula Weight 179.17
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tricine [Good′s buffer component for biological research]