missing translation for 'onlineSavingsMsg'
Learn More

Trichloromelamine 95.0+%, TCI America™

Catalog No. p-7136250
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
T038425G 25 g
T0384100G 100 g
2 options

Catalog No. T038425G

Supplier: TCI America T038425G

Only null left

Chemical Identifiers

CAS 7673-09-8
Molecular Formula C3H3Cl3N6
Molecular Weight (g/mol) 229.45
MDL Number MFCD00006047
InChI Key KEPNSIARSTUPGS-UHFFFAOYSA-N
Synonym 2,4,6-Tris(chloroamino)-1,3,5-triazine
PubChem CID 24322
IUPAC Name 2-N,4-N,6-N-trichloro-1,3,5-triazine-2,4,6-triamine
SMILES C1(=NC(=NC(=N1)NCl)NCl)NCl

Specifications

Color Yellow
Quantity 25 g
UN Number 1479
Formula Weight 229.45
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trichloromelamine
RTECSNumber XZ1575000
TSCA Yes
Recommended Storage Refrigerator