missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroisocyanuric Acid 95.0+%, TCI America™
Supplier: TCI America T0620500G
Specifications
| Trichloroisocyanuric Acid | |
| White | |
| MFCD00006553 | |
| 2468 | |
| YRIZYWQGELRKNT-UHFFFAOYSA-N | |
| trichloro-1,3,5-triazinane-2,4,6-trione | |
| 6909 | |
| 232.40 | |
| Crystalline Powder |
| 87-90-1 | |
| C3Cl3N3O3 | |
| 500 g | |
| trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal | |
| ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O | |
| 232.40 | |
| CHEBI:33015 | |
| ≥95.0% (T) |
Chemical Identifiers
| 87-90-1 | |
| 232.40 | |
| YRIZYWQGELRKNT-UHFFFAOYSA-N | |
| 6909 | |
| trichloro-1,3,5-triazinane-2,4,6-trione |
| C3Cl3N3O3 | |
| MFCD00006553 | |
| trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal | |
| CHEBI:33015 | |
| ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O |
Safety and Handling
EINECSNumber : (5)-1044
RTECSNumber : XZ1925000
TSCA : Yes