missing translation for 'onlineSavingsMsg'
Learn More

Trichloroisocyanuric Acid 95.0+%, TCI America™

Catalog No. T062025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T062025G 25 g
T0620500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T062025G Supplier TCI America Supplier No. T062025G

Chemical Identifiers

CAS 87-90-1
Molecular Formula C3Cl3N3O3
Molecular Weight (g/mol) 232.40
MDL Number MFCD00006553
InChI Key YRIZYWQGELRKNT-UHFFFAOYSA-N
Synonym trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal
PubChem CID 6909
ChEBI CHEBI:33015
IUPAC Name trichloro-1,3,5-triazinane-2,4,6-trione
SMILES ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O

Specifications

Color White
Quantity 25 g
UN Number 2468
Formula Weight 232.40
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trichloroisocyanuric Acid
EINECSNumber (5)-1044
RTECSNumber XZ1925000
TSCA Yes