missing translation for 'onlineSavingsMsg'
Learn More

Trichloroisocyanuric acid, 90+%

Catalog No. AAB2390622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2390622 100 g
AAB2390636 500 g
AAB239060E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2390622 Supplier Thermo Scientific Chemicals Supplier No. B2390622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.

Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).

Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.

Chemical Identifiers

CAS 87-90-1
Molecular Formula C3Cl3N3O3
Molecular Weight (g/mol) 232.40
MDL Number MFCD00006553
InChI Key YRIZYWQGELRKNT-UHFFFAOYSA-N
Synonym trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal
PubChem CID 6909
ChEBI CHEBI:33015
SMILES ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O

Specifications

Melting Point ∼245°C (decomposition)
Density 2.07
pH 3
Flash Point >250°C (482°F)
Odor Pungent
Quantity 100 g
UN Number UN2468
Beilstein 202022
Sensitivity Moisture sensitive
Merck Index 14,9641
Solubility Information Soluble in chlorocarbon,acetonitrile,acetone,water (12g/L).
IUPAC Name trichloro-1,3,5-triazinane-2,4,6-trione
Formula Weight 232.41
Percent Purity ≥90%
Chemical Name or Material Trichloroisocyanuric acid
Show More Show Less
Hazard Category H272-H302-H319-H335
Hazard Statement GHS H Statement
H272-H302-H319-H335
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: TRICHLOROISOCYANURIC ACID, DRY
EINECSNumber 201-782-8
RTECSNumber XZ1925000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only