missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, Purified (Trichloroethene), Reagents

Catalog No. 01336913
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336913 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336913 Supplier Labchem Inc Supplier No. C234200020E6
Only null left

Trichloroethylene, Purified (Trichloroethene), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym Trichloroethene, 1,1,2-Trichloroethene
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

Boiling Range 87°C
Melting Point -84.8°C
Color Colorless
Boiling Point 87°C
Melting Range -84.8°C
Packaging Steel Pail
Quantity 20 L
Specific Gravity 1.47
Grade Reagent
Physical Form Liquid
Chemical Name or Material Trichloroethylene, Purified (Trichloroethene)
Show More Show Less