missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, Purified (Trichloroethene), Reagents

Catalog No. 01336912
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
20 L
4 L
Catalog No. Quantity
01336912 4 L
01336913 20 L
2 options

Catalog No. 01336912

Supplier: Labchem Inc C23420004C

Only null left

Trichloroethylene, Purified (Trichloroethene), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym Trichloroethene, 1,1,2-Trichloroethene
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

Boiling Range 87°C
Melting Point -84.8°C
Color Colorless
Boiling Point 87°C
Melting Range -84.8°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 1.47
Grade Reagent
Physical Form Liquid
Chemical Name or Material Trichloroethylene, Purified (Trichloroethene)
Show More Show Less