missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, ACS Reagent (Trichloroethene), Reagents

Catalog No. 01337640
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337640 20 L
01337639 4 L
01337638 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337640 Supplier Labchem Inc Supplier No. C134200020E6
Only null left

Trichloroethylene, ACS Reagent (Trichloroethene), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym Trichloroethene, 1,1,2-Trichloroethene
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

Boiling Range 87°C
Melting Point -84.8°C
Color Colorless
Boiling Point 87°C
Melting Range -84.8°C
Packaging Steel Pail
Quantity 20 L
Specific Gravity 1.47
Grade ACS
Physical Form Liquid
Chemical Name or Material Trichloroethylene, ACS Reagent (Trichloroethene)
Show More Show Less