missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, ACS Reagent (Trichloroethene), Reagents

Catalog No. 01337638
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337638 500 mL
01337640 20 L
01337639 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337638 Supplier Labchem Inc Supplier No. C1342000500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trichloroethylene, ACS Reagent (Trichloroethene), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym Trichloroethene, 1,1,2-Trichloroethene
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

Boiling Range 87°C
Melting Point -84.8°C
Color Colorless
Boiling Point 87°C
Melting Range -84.8°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 1.47
Grade ACS
Physical Form Liquid
Chemical Name or Material Trichloroethylene, ACS Reagent (Trichloroethene)
Show More Show Less