missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, ACS, 99.5%, Spectrum™ Chemical

Catalog No. p-7166655
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614527 500 mL Amber Glass Bottle
18614524 4 L Amber Glass Bottle
18614525 20 L Steel Can
18614526 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614527 Supplier Spectrum Chemical Mfg Cor Supplier No. T1115500MLGL
Only null left

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Residue after Evaporation 1e-05
Percent Purity 0.995
Grade ACS
Titratable Acid 0.0001 meq/g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.