missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, 99+%, extra pure, stabilized

Catalog No. p-3737642
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158310025 2.5 L Glass Bottle
AC158310250 25 L Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158310025 Supplier Thermo Scientific Chemicals Supplier No. 158310025
Only null left

CAS: 79-01-6 | C2HCl3 | 131.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-01-6
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl

Specifications

Melting Point -85°C
Density 1.4600g/mL
Boiling Point 87.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Molecular Formula C2HCl3
Linear Formula ClCH=CCl2
Refractive Index 1.4760 to 1.4780
Quantity 2.5 L
Beilstein 01,187
Fieser 09,479; 10,414; 11,552
Merck Index 15,9804
Specific Gravity 1.46
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Molecular Weight (g/mol) 131.39
Viscosity 0.55 mPa.s (25°C)
Formula Weight 131.39
Vapor Pressure 77.3mbar at 20°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Trichloroethylene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
EINECSNumber 201-167-4

RUO – Research Use Only