missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, 99.6%, ACS reagent, stabilized

Catalog No. AC421520025
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421520025 2.5 L Glass bottle
AC421525000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421520025 Supplier Thermo Scientific Chemicals Supplier No. 421520025
Only null left

CAS: 79-01-6 | C2HCl3 | 131.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.39
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl

Specifications

Melting Point -85°C
Density 1.4600g/mL
Boiling Point 87.0°C
Assay Percent Range 99.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH=CCl2
Refractive Index 1.4760 to 1.4780
Quantity 2.5 L
Residue after Evaporation 0.001% max.
Beilstein 01, 187
Fieser 09,479; 10,414; 11,552
Specific Gravity 1.46
Merck Index 15, 9804
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Stabilizer 0.5% (typically) Ethanol, or 0.8% (typically) Ethyloxirane
Viscosity 0.55 mPa.s (25°C)
Formula Weight 131.39
Percent Purity 99.6%
Grade ACS Reagent
Titratable Acid 0.0001 meq/g max
Physical Form Liquid
Chemical Name or Material Trichloroethylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
EINECSNumber 201-167-4
RTECSNumber KX4550000
TSCA TSCA

RUO – Research Use Only