missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, 98%

CAS: 79-01-6 | C2HCl3 | 131.38 g/mol

Supplier:  Thermo Scientific Chemicals L144740F

 View more versions of this product

Catalog No. AAL144740F

Only null left
Explore available promotions

Description

Description

Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane

Solubility
Soluble in water (1g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light Sensitive to light. Handle and store under inert gas. Incompatible with bases and oxidizing agents.
Specifications

Specifications

Trichloroethylene
-85°C
87°C
C2HCl3
MFCD00000838
UN1710
14,9639
Soluble in water (1g/L at 20°C).
C(=C(Cl)Cl)Cl
131.38
CHEBI:16602
98%
79-01-6
1.46
Chloroform-like
1.476
2500 mL
1736782
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
XSTXAVWGXDQKEL-UHFFFAOYSA-N
1,1,2-trichloroethene
6575
131.39

Chemical Identifiers

79-01-6
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
6575
1,1,2-trichloroethene
C2HCl3
MFCD00000838
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
CHEBI:16602
C(=C(Cl)Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c

H315-H317-H319-H336-H341-H350-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE

EINECSNumber : 201-167-4

RTECSNumber : KX4550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only