missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, 98%

Catalog No. AAL14474AE
Change view
Click to view available options
Quantity:
100 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14474AE 100 mL
AAL144740F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14474AE Supplier Thermo Scientific Chemicals Supplier No. L14474AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-01-6 | C2HCl3 | 131.38 g/mol

Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroethylene is used as household cleaner, with trichloroethane it is used in most typewriter correction fluid. It is also used in wool-fabric scouring, as extractant for spice oleoresins. Intermediate in the production of pentachloroethane

Solubility
Soluble in water (1g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light Sensitive to light. Handle and store under inert gas. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl

Specifications

Melting Point -85°C
Density 1.46
Boiling Point 87°C
Odor Chloroform-like
Refractive Index 1.476
Quantity 100 mL
UN Number UN1710
Beilstein 1736782
Merck Index 14,9639
Solubility Information Soluble in water (1g/L at 20°C).
Formula Weight 131.39
Percent Purity 98%
Chemical Name or Material Trichloroethylene
Show More Show Less
Hazard Category H315-H317-H319-H336-H341-H350-H373
Hazard Statement GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE
EINECSNumber 201-167-4
RTECSNumber KX4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only