missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetyl Isocyanate 97.0+%, TCI America™

Catalog No. T10475G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10475G 5 g
T10471G 1 g
T104725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T10475G Supplier TCI America Supplier No. T10475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3019-71-4
Molecular Formula C3Cl3NO2
Molecular Weight (g/mol) 188.39
MDL Number MFCD00002033
InChI Key GRNOZCCBOFGDCL-UHFFFAOYSA-N
Synonym trichloroacetyl isocyanate, acetyl isocyanate, trichloro, trichloroacetylisocyanate, isocyanic acid trichloroacetyl ester, unii-qxv1p5j6jl, qxv1p5j6jl, acetyl isocyanate, 2,2,2-trichloro, 2,2,2-trichloroethanecarbonyl isocyanate, trichloroethanecarbonyl isocyanate, trichloroacetyl isocyanate, nmr grade
PubChem CID 76400
IUPAC Name trichloroethanecarbonyl isocyanate
SMILES ClC(Cl)(Cl)C(=O)N=C=O

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 2206
Formula Weight 188.39
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material Trichloroacetyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator